CAS RN 6335-39-3
IUPAC Name:
4-(propan-2-yl)benzene-1-sulfonamide
Synonyms:
4-isopropylbenzenesulfonamide
Molecular Weight (g/mol):
199.27
Molecular Formula:
C9H13NO2S
InChi Key:
WVOWEROKBOQYLJ-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=C(C=C1)S(N)(=O)=O
1
–
1
of
1
results
1
Promotions Available
4-Isopropylbenzenesulfonamide, 97%
4-Isopropylbenzenesulfonamide, 97%
CAS: 6335-39-3 Molecular Formula: C9H13NO2S Molecular Weight (g/mol): 199.268 MDL Number: MFCD00457071 InChI Key: WVOWEROKBOQYLJ-UHFFFAOYSA-N Synonym: 4-isopropylbenzenesulfonamide,p-cumenesulfonamide,4-1-methylethyl benzenesulfonamide,4-propan-2-yl benzene-1-sulfonamide,4-i-propyl benzenesulfonamide,4-methylethyl benzenesulfonamide,p-isopropylbenzenesulfonamide,4-isopropylbenzenesulphonamide,4-iso-propylbenzenesulfonamide,4-isopropyl-benzenesulfonamide PubChem CID: 232314 IUPAC Name: 4-propan-2-ylbenzenesulfonamide SMILES: CC(C)C1=CC=C(C=C1)S(=O)(=O)N