missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isopropoxybenzoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 163°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Synonym | 4-isopropoxybenzoic acid, benzoic acid, 4-1-methylethoxy, 4-propan-2-yloxy benzoic acid, 4-isopropoxybenzoicacid, 4-isopropoxy-benzoic acid, 4-iso-propyloxybenzoic acid, rarechem al be 0294, 4-isopropyloxybenzoic acid, 4-1-methylethoxy benzoic acid, benzoic acid,4-1-methylethoxy |
| Formula Weight | 180.20 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Isopropoxybenzoic Acid |
Safety and Handling
| TSCA | No |