accessibility menu, dialog, popup

UserName

CAS RN 13205-46-4

IUPAC Name: 4-(propan-2-yloxy)benzoic acid
Synonyms: 4-isopropoxybenzoic acid
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: ZVERWTXKKWSSHH-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)