missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropoxybenzoic acid, 99%

Catalog No. AAA1435130
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1435130 250 g
AAA1435109 10 g
AAA1435118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1435130 Supplier Thermo Scientific Chemicals Supplier No. A1435130

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13205-46-4 | C10H12O3 | 180.203 g/mol

4-Isopropoxybenzoic acid is used to prepare 5-(4-isopropoxyphenyl)-N-phenyl-1,3,4-thiadiazol-2-amine by reacting with N-phenylhydrazinecarbothioamide in the presence of phosphorusoxychloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Isopropoxybenzoic acid is used to prepare 5-(4-isopropoxyphenyl)-N-phenyl-1,3,4-thiadiazol-2-amine by reacting with N-phenylhydrazinecarbothioamide in the presence of phosphorusoxychloride.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 13205-46-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00044318
InChI Key ZVERWTXKKWSSHH-UHFFFAOYSA-N
PubChem CID 72972
IUPAC Name 4-propan-2-yloxybenzoic acid
SMILES CC(C)OC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 163°C to 167°C
Odor Odorless
Quantity 250 g
Synonym 4-isopropoxybenzoic acid, benzoic acid, 4-1-methylethoxy, 4-propan-2-yloxy benzoic acid, 4-isopropoxybenzoicacid, 4-isopropoxy-benzoic acid, 4-iso-propyloxybenzoic acid, rarechem al be 0294, 4-isopropyloxybenzoic acid, 4-1-methylethoxy benzoic acid, benzoic acid,4-1-methylethoxy
Solubility Information Insoluble in water.
Formula Weight 180.2
Percent Purity 99%
Chemical Name or Material 4-Isopropoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-169-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only