missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropenyltoluene (stabilized with TBC) 95.0+%, TCI America™

Catalog No. p-7131546
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06935G 5 g
I069325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06935G Supplier TCI America Supplier No. I06935G
Only null left

Chemical Identifiers

CAS 1195-32-0
Molecular Formula C10H12
Molecular Weight (g/mol) 132.21
MDL Number MFCD00036510
InChI Key MMSLOZQEMPDGPI-UHFFFAOYSA-N
Synonym 1-methyl-4-prop-1-en-2-yl benzene, dehydro-p-cymene, 2-p-tolylpropene, 4-isopropenyltoluene, 4-methylisopropenylbenzene, 2-p-methylphenyl propene, p-cymenene, p,alpha-dimethylstyrene, p-alpha-dimethylstyrene, 1-isopropenyl-4-methylbenzene
PubChem CID 62385
IUPAC Name 1-methyl-4-(prop-1-en-2-yl)benzene
SMILES CC(=C)C1=CC=C(C)C=C1

Specifications

Melting Point -28°C
Color Yellow
Boiling Point 186°C
Quantity 5 g
Formula Weight 132.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Isopropenyltoluene (stabilized with TBC)
EINECSNumber (3)-0008&(3)-0010
TSCA Yes
Recommended Storage Freezer