missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isobutylbenzoic Acid 99.0+%, TCI America™
$397.05 - $740.03
Chemical Identifiers
| CAS | 38861-88-0 |
|---|---|
| Molecular Formula | C11H14O2 |
| Molecular Weight (g/mol) | 178.23 |
| MDL Number | MFCD00191658 |
| InChI Key | VUBBCFWWSKOHTH-UHFFFAOYSA-N |
| PubChem CID | 38111 |
| IUPAC Name | 4-(2-methylpropyl)benzoic acid |
| SMILES | CC(C)CC1=CC=C(C=C1)C(O)=O |
Chemical Identifiers
| 38861-88-0 | |
| 178.23 | |
| VUBBCFWWSKOHTH-UHFFFAOYSA-N | |
| 4-(2-methylpropyl)benzoic acid |
| C11H14O2 | |
| MFCD00191658 | |
| 38111 | |
| CC(C)CC1=CC=C(C=C1)C(O)=O |
Specifications
| 38861-88-0 | |
| White | |
| MFCD00191658 | |
| VUBBCFWWSKOHTH-UHFFFAOYSA-N | |
| 4-(2-methylpropyl)benzoic acid | |
| 38111 | |
| ≥99.0% (GC,T) | |
| 4-Isobutylbenzoic Acid |
| 144°C | |
| C11H14O2 | |
| 10 g | |
| CC(C)CC1=CC=C(C=C1)C(O)=O | |
| 178.23 | |
| 178.23 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1338
RTECSNumber : DH3073800
TSCA : No