missing translation for 'onlineSavingsMsg'
Learn More

4-Isobutylbenzoic Acid 99.0+%, TCI America™

Catalog No. i043725g
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I043725G 25 g
I043710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I043725G Supplier TCI America Supplier No. I043725G
Only null left

Chemical Identifiers

CAS 38861-88-0
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00191658
InChI Key VUBBCFWWSKOHTH-UHFFFAOYSA-N
PubChem CID 38111
IUPAC Name 4-(2-methylpropyl)benzoic acid
SMILES CC(C)CC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 144°C
Color White
Quantity 25 g
Formula Weight 178.23
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Isobutylbenzoic Acid
EINECSNumber (3)-1338
RTECSNumber DH3073800
TSCA No