missing translation for 'onlineSavingsMsg'
Learn More

4-Isobutylbenzaldehyde 95.0+%, TCI America™

Catalog No. I0436500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0436500ML 500 mL
I043625ML 25 mL
I0436100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0436500ML Supplier TCI America Supplier No. I0436500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40150-98-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.232
MDL Number MFCD00075760
InChI Key LXPWGAZYJHUWPM-UHFFFAOYSA-N
PubChem CID 577537
IUPAC Name 4-(2-methylpropyl)benzaldehyde
SMILES CC(C)CC1=CC=C(C=C1)C=O

Specifications

Color Yellow
Quantity 500 mL
Formula Weight 162.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Isobutylbenzaldehyde
EINECSNumber (3)-2663
TSCA Yes