CAS RN 40150-98-9
IUPAC Name:
4-(2-methylpropyl)benzaldehyde
Synonyms:
4-(2-methylpropyl)benzaldehyde
Molecular Weight (g/mol):
162.23
Molecular Formula:
C11H14O
InChi Key:
LXPWGAZYJHUWPM-UHFFFAOYSA-N
SMILES:
CC(C)CC1=CC=C(C=O)C=C1
1
–
1
of
1
results
1
4-Isobutylbenzaldehyde 95.0+%, TCI America™
CAS: 40150-98-9 Molecular Formula: C11H14O Molecular Weight (g/mol): 162.232 MDL Number: MFCD00075760 InChI Key: LXPWGAZYJHUWPM-UHFFFAOYSA-N PubChem CID: 577537 IUPAC Name: 4-(2-methylpropyl)benzaldehyde SMILES: CC(C)CC1=CC=C(C=C1)C=O