missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenylsulfur Pentafluoride 94.0+%, TCI America™

Catalog No. I06951G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06951G 1 g
I06955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06951G Supplier TCI America Supplier No. I06951G
Only null left

Chemical Identifiers

CAS 286947-68-0
Molecular Formula C6H4F5IS
Molecular Weight (g/mol) 330.05
MDL Number MFCD03788514
InChI Key FRYANWYSCROOCU-UHFFFAOYSA-N
PubChem CID 2779199
IUPAC Name 1-iodo-4-(pentafluoro-λ⁶-sulfanyl)benzene
SMILES FS(F)(F)(F)(F)C1=CC=C(I)C=C1

Specifications

Melting Point 36°C
Color Yellow
Boiling Point 52°C
Quantity 1 g
Formula Weight 330.05
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodophenylsulfur Pentafluoride
TSCA No
Recommended Storage Refrigerator