accessibility menu, dialog, popup

UserName

CAS RN 286947-68-0

SFFFFFI

CAS RN 286947-68-0

IUPAC Name: 1-iodo-4-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms: 1-iodo-4-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol): 330.05
Molecular Formula: C6H4F5IS
InChi Key: FRYANWYSCROOCU-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC=C(I)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results