missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenylhydrazine, 95%, Thermo Scientific™

$97.74 - $358.51

Chemical Identifiers

CAS 13116-27-3
Molecular Formula C6H7IN2
Molecular Weight (g/mol) 234.04
MDL Number MFCD00052688
InChI Key IQMLCMKMSBMMGR-UHFFFAOYSA-N
Synonym 4-iodophenyl hydrazine, 1-4-iodophenyl hydrazine, 4-iodo-phenyl-hydrazine, 4-iodo phenyl hydrazine, p-iodophenyl-hydrazine, 4-iodophenyl diazane, pubchem15797, acmc-1brf1, 1-hydrazino-4-iodobenzene, 4-iodophenylhydrazine
PubChem CID 704591
IUPAC Name (4-iodophenyl)hydrazine
SMILES C1=CC(=CC=C1NN)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC399050010
View Documents Promotion Details
N/A 1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC399050050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13116-27-3
234.04
IQMLCMKMSBMMGR-UHFFFAOYSA-N
704591
C1=CC(=CC=C1NN)I
C6H7IN2
MFCD00052688
4-iodophenyl hydrazine, 1-4-iodophenyl hydrazine, 4-iodo-phenyl-hydrazine, 4-iodo phenyl hydrazine, p-iodophenyl-hydrazine, 4-iodophenyl diazane, pubchem15797, acmc-1brf1, 1-hydrazino-4-iodobenzene, 4-iodophenylhydrazine
(4-iodophenyl)hydrazine

Specifications

13116-27-3
100.0
C6H7IN2
MFCD00052688
Solubility in water: insoluble
C1=CC(=CC=C1NN)I
234.04
234.04
4-Iodophenylhydrazine
94.0
95%
IC6H4NHNH2
4-iodophenyl hydrazine, 1-4-iodophenyl hydrazine, 4-iodo-phenyl-hydrazine, 4-iodo phenyl hydrazine, p-iodophenyl-hydrazine, 4-iodophenyl diazane, pubchem15797, acmc-1brf1, 1-hydrazino-4-iodobenzene, 4-iodophenylhydrazine
IQMLCMKMSBMMGR-UHFFFAOYSA-N
(4-iodophenyl)hydrazine
704591
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause an allergic skin reaction.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.<br/

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fre

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only