Learn More
4-Iodophenylhydrazine, 95%
CAS: 13116-27-3 | C6H7IN2 | 234.04 g/mol
Supplier: Thermo Scientific Chemicals L1480606
Description
It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water.
Notes
Light Sensitive, store in dark. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 4-Iodophenylhydrazine | |
| 99°C to 103°C | |
| MFCD00052688 | |
| 742475 | |
| 4-iodophenyl hydrazine, 1-4-iodophenyl hydrazine, 4-iodo-phenyl-hydrazine, 4-iodo phenyl hydrazine, p-iodophenyl-hydrazine, 4-iodophenyl diazane, pubchem15797, acmc-1brf1, 1-hydrazino-4-iodobenzene, 4-iodophenylhydrazine | |
| IQMLCMKMSBMMGR-UHFFFAOYSA-N | |
| (4-iodophenyl)hydrazine | |
| 704591 | |
| 95% |
| 13116-27-3 | |
| C6H7IN2 | |
| 5 g | |
| Light sensitive | |
| Insoluble in water. | |
| C1=CC(=CC=C1NN)I | |
| 234.04 | |
| 234.04 |
Chemical Identifiers
| 13116-27-3 | |
| 234.04 | |
| IQMLCMKMSBMMGR-UHFFFAOYSA-N | |
| 704591 | |
| C1=CC(=CC=C1NN)I |
| C6H7IN2 | |
| MFCD00052688 | |
| 4-iodophenyl hydrazine, 1-4-iodophenyl hydrazine, 4-iodo-phenyl-hydrazine, 4-iodo phenyl hydrazine, p-iodophenyl-hydrazine, 4-iodophenyl diazane, pubchem15797, acmc-1brf1, 1-hydrazino-4-iodobenzene, 4-iodophenylhydrazine | |
| (4-iodophenyl)hydrazine |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H315-H317-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only