missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodophenyl isocyanate, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 43°C to 47°C |
| Boiling Point | 84°C (5 mmHg) |
| Flash Point | >110°C (230°F) |
| Odor | Pungent |
| Quantity | 1 g |
| UN Number | UN2811 |
| Beilstein | 2716151 |
| Sensitivity | Light and moisture sensitive |
| Formula Weight | 245.02 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H317-H319-H331-H335 |
| Hazard Statement | GHS H Statement H331-H334-H302-H315-H319-H335 Toxic if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P333+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only