accessibility menu, dialog, popup

UserName

CAS RN 15845-62-2

IUPAC Name: 1-iodo-4-isocyanatobenzene
Synonyms: 1-iodo-4-isocyanatobenzene
Molecular Weight (g/mol): 245.02
Molecular Formula: C7H4INO
InChi Key: KVMKUTIUJGYHBA-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=C1)N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results