missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenyl isocyanate, 97%

Catalog No. AAL1600003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1600003 1 g
AAL1600006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1600003 Supplier Thermo Scientific Chemicals Supplier No. L1600003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 15845-62-2 | C7H4INO | 245.019 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15845-62-2
Molecular Formula C7H4INO
Molecular Weight (g/mol) 245.019
MDL Number MFCD00013875
InChI Key KVMKUTIUJGYHBA-UHFFFAOYSA-N
Synonym 4-iodophenyl isocyanate, 4-iodophenylisocyanate, 1-iodo-4-isocyanato-benzene, p-iodophenylisocyanate, acmc-1bsbj, benzene,1-iodo-4-isocyanato
PubChem CID 3399787
IUPAC Name 1-iodo-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)I

Specifications

Melting Point 43°C to 47°C
Boiling Point 84°C (5 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Quantity 1 g
UN Number UN2811
Beilstein 2716151
Sensitivity Light and moisture sensitive
Formula Weight 245.02
Percent Purity 97%
Chemical Name or Material 4-Iodophenyl isocyanate
Show More Show Less
Hazard Category H302-H315-H317-H319-H331-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only