missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenol 98.0+%, TCI America™

Catalog No. I031110G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I031110G 10 g
I031125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I031110G Supplier TCI America Supplier No. I031110G

Chemical Identifiers

CAS 540-38-5
Molecular Formula C6H5IO
Molecular Weight (g/mol) 220.01
MDL Number MFCD00002327
InChI Key VSMDINRNYYEDRN-UHFFFAOYSA-N
Synonym p-iodophenol, phenol, 4-iodo, phenol, p-iodo, 4-hydroxyiodobenzene, p-hydroxyiodobenzene, 4-hydroxyphenyl iodide, 4-iodo-phenol, unii-bh194bak0b, ccris 668, bh194bak0b
PubChem CID 10894
ChEBI CHEBI:43521
IUPAC Name 4-iodophenol
SMILES OC1=CC=C(I)C=C1

Specifications

Melting Point 91°C
Quantity 10 g
Formula Weight 220.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodophenol
RTECSNumber SL5600000
TSCA Yes