missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 4-Iodophenol [for Biochemical Research] | |
| 93°C | |
| MFCD00002327 | |
| p-iodophenol, phenol, 4-iodo, phenol, p-iodo, 4-hydroxyiodobenzene, p-hydroxyiodobenzene, 4-hydroxyphenyl iodide, 4-iodo-phenol, unii-bh194bak0b, ccris 668, bh194bak0b | |
| OC1=CC=C(I)C=C1 | |
| 220.01 | |
| CHEBI:43521 | |
| ≥98.0% (GC) |
| 540-38-5 | |
| C6H5IO | |
| 5 g | |
| VSMDINRNYYEDRN-UHFFFAOYSA-N | |
| 4-iodophenol | |
| 10894 | |
| 220.01 | |
| Crystalline Powder |
Chemical Identifiers
| 540-38-5 | |
| 220.01 | |
| VSMDINRNYYEDRN-UHFFFAOYSA-N | |
| 10894 | |
| 4-iodophenol |
| C6H5IO | |
| MFCD00002327 | |
| p-iodophenol, phenol, 4-iodo, phenol, p-iodo, 4-hydroxyiodobenzene, p-hydroxyiodobenzene, 4-hydroxyphenyl iodide, 4-iodo-phenol, unii-bh194bak0b, ccris 668, bh194bak0b | |
| CHEBI:43521 | |
| OC1=CC=C(I)C=C1 |
Safety and Handling
RTECSNumber : SL5600000
TSCA : Yes