missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzylamine 97.0+%, TCI America™

Catalog No. I08311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08311G 1 g
I08315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08311G Supplier TCI America Supplier No. I08311G

Chemical Identifiers

CAS 39959-59-6
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.052
MDL Number MFCD00047933
InChI Key KCGZGJOBKAXVSU-UHFFFAOYSA-N
Synonym 4-iodophenyl methanamine, 4-iodobenzylamine, 4-iodo-benzylamine, 4-iodobenzyl amine, benzenemethanamine, 4-iodo, 1-4-iodophenyl methanamine, 4-iodophenyl methylamine, 4-iodobenzylamine, hydrochloride salt, acmc-20a5sy, p-iodobenzylamine
PubChem CID 2737533
IUPAC Name (4-iodophenyl)methanamine
SMILES C1=CC(=CC=C1CN)I

Specifications

Melting Point 48°C
Boiling Point 115°C
Quantity 1 g
UN Number 3259
Formula Weight 233.05
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodobenzylamine