accessibility menu, dialog, popup

UserName

CAS RN 39959-59-6

IUPAC Name: 1-(4-iodophenyl)methanamine
Synonyms: 1-(4-iodophenyl)methanamine
Molecular Weight (g/mol): 233.05
Molecular Formula: C7H8IN
InChi Key: KCGZGJOBKAXVSU-UHFFFAOYSA-N
SMILES: NCC1=CC=C(I)C=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)