missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzylamine, 97%

Catalog No. p-7047046
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1745003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1745003 Supplier Thermo Scientific Chemicals Supplier No. L1745003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39959-59-6 | C7H8IN | 233.052 g/mol

4-Iodobenzylamine is used to prepare 2-bromo-N-(4-iodo-benzyl)-acetamide in the presence of bromoacetyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzylamine is used to prepare 2-bromo-N-(4-iodo-benzyl)-acetamide in the presence of bromoacetyl bromide.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air and light. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 39959-59-6
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.052
MDL Number MFCD00047933
InChI Key KCGZGJOBKAXVSU-UHFFFAOYSA-N
Synonym 4-iodophenyl methanamine, 4-iodobenzylamine, 4-iodo-benzylamine, 4-iodobenzyl amine, benzenemethanamine, 4-iodo, 1-4-iodophenyl methanamine, 4-iodophenyl methylamine, 4-iodobenzylamine, hydrochloride salt, acmc-20a5sy, p-iodobenzylamine
PubChem CID 2737533
IUPAC Name (4-iodophenyl)methanamine
SMILES C1=CC(=CC=C1CN)I

Specifications

Melting Point 46°C to 48°C
Boiling Point 113°C to 115°C (4 mmHg)
Quantity 1 g
UN Number UN3259
Beilstein 2689605
Sensitivity Air and light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 233.05
Percent Purity 97%
Chemical Name or Material 4-Iodobenzylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only