missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodobenzyl Bromide 97.0+%, TCI America™
$115.57 - $516.35
Chemical Identifiers
| CAS | 16004-15-2 |
|---|---|
| Molecular Formula | C7H6BrI |
| Molecular Weight (g/mol) | 296.933 |
| MDL Number | MFCD00209656 |
| InChI Key | BQTRMYJYYNQQGK-UHFFFAOYSA-N |
| Synonym | 4-iodobenzyl bromide, 1-bromomethyl-4-iodobenzene, 4-iodobenzylbromide, p-iodobenzyl bromide, 1-bromomethyl-4-iodo-benzene, alpha-bromo-4-iodotoluene, 4-bromomethyl-1-iodobenzene, benzene, 1-bromomethyl-4-iodo, 4-iodo benzyl bromide |
| PubChem CID | 954258 |
| IUPAC Name | 1-(bromomethyl)-4-iodobenzene |
| SMILES | C1=CC(=CC=C1CBr)I |
Chemical Identifiers
| 16004-15-2 | |
| 296.933 | |
| BQTRMYJYYNQQGK-UHFFFAOYSA-N | |
| 954258 | |
| C1=CC(=CC=C1CBr)I |
| C7H6BrI | |
| MFCD00209656 | |
| 4-iodobenzyl bromide, 1-bromomethyl-4-iodobenzene, 4-iodobenzylbromide, p-iodobenzyl bromide, 1-bromomethyl-4-iodo-benzene, alpha-bromo-4-iodotoluene, 4-bromomethyl-1-iodobenzene, benzene, 1-bromomethyl-4-iodo, 4-iodo benzyl bromide | |
| 1-(bromomethyl)-4-iodobenzene |
Specifications
| 16004-15-2 | |
| White-Yellow | |
| MFCD00209656 | |
| 3261 | |
| BQTRMYJYYNQQGK-UHFFFAOYSA-N | |
| 1-(bromomethyl)-4-iodobenzene | |
| 954258 | |
| ≥97.0% (GC) | |
| 4-Iodobenzyl Bromide |
| 80°C | |
| C7H6BrI | |
| 5 g | |
| 4-iodobenzyl bromide, 1-bromomethyl-4-iodobenzene, 4-iodobenzylbromide, p-iodobenzyl bromide, 1-bromomethyl-4-iodo-benzene, alpha-bromo-4-iodotoluene, 4-bromomethyl-1-iodobenzene, benzene, 1-bromomethyl-4-iodo, 4-iodo benzyl bromide | |
| C1=CC(=CC=C1CBr)I | |
| 296.933 | |
| 296.93 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator