accessibility menu, dialog, popup

UserName

CAS RN 16004-15-2

IUPAC Name: 1-(bromomethyl)-4-iodobenzene
Synonyms: 1-(bromomethyl)-4-iodobenzene
Molecular Weight (g/mol): 296.93
Molecular Formula: C7H6BrI
InChi Key: BQTRMYJYYNQQGK-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(I)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)