missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzyl Bromide 97.0+%, TCI America™

Catalog No. I05715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05715G 5 g
I057125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05715G Supplier TCI America Supplier No. I05715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16004-15-2
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.933
MDL Number MFCD00209656
InChI Key BQTRMYJYYNQQGK-UHFFFAOYSA-N
Synonym 4-iodobenzyl bromide, 1-bromomethyl-4-iodobenzene, 4-iodobenzylbromide, p-iodobenzyl bromide, 1-bromomethyl-4-iodo-benzene, alpha-bromo-4-iodotoluene, 4-bromomethyl-1-iodobenzene, benzene, 1-bromomethyl-4-iodo, 4-iodo benzyl bromide
PubChem CID 954258
IUPAC Name 1-(bromomethyl)-4-iodobenzene
SMILES C1=CC(=CC=C1CBr)I

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 296.93
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodobenzyl Bromide
TSCA No
Recommended Storage Refrigerator