missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzyl bromide, 97%

Catalog No. AAA1820006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1820006 5 g
AAA1820014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1820006 Supplier Thermo Scientific Chemicals Supplier No. A1820006

CAS: 16004-15-2 | C7H6BrI | 296.933 g/mol

4-Iodobenzyl bromide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzyl bromide is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents, bases and amines.

Chemical Identifiers

CAS 16004-15-2
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.933
MDL Number MFCD00209656
InChI Key BQTRMYJYYNQQGK-UHFFFAOYSA-N
Synonym 4-iodobenzyl bromide, 1-bromomethyl-4-iodobenzene, 4-iodobenzylbromide, p-iodobenzyl bromide, 1-bromomethyl-4-iodo-benzene, alpha-bromo-4-iodotoluene, 4-bromomethyl-1-iodobenzene, benzene, 1-bromomethyl-4-iodo, 4-iodo benzyl bromide
PubChem CID 954258
IUPAC Name 1-(bromomethyl)-4-iodobenzene
SMILES C1=CC(=CC=C1CBr)I

Specifications

Melting Point 78°C to 82°C
Boiling Point 140°C (15 mmHg)
Odor Irritating
Quantity 5 g
UN Number UN3261
Beilstein 2325160
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 296.94
Percent Purity 97%
Chemical Name or Material 4-Iodobenzyl bromide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only