missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzyl Alcohol 99.0+%, TCI America™

Catalog No. p-7131606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08105G 5 g
I081025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08105G Supplier TCI America Supplier No. I08105G
Only null left

Chemical Identifiers

CAS 18282-51-4
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD01732720
InChI Key CNQRHSZYVFYOIE-UHFFFAOYSA-N
Synonym 4-iodobenzyl alcohol, 4-iodophenyl methanol, p-iodobenzyl alcohol, benzenemethanol, 4-iodo, benzyl alcohol, p-iodo, 4-iodobenzylalcohol, p-iodobenzylalcohol, 4-iodobenzyl-alcohol, 4-iodobenzyl zlcohol, p-lodo-benzyl alcohol
PubChem CID 29012
IUPAC Name (4-iodophenyl)methanol
SMILES C1=CC(=CC=C1CO)I

Specifications

Melting Point 73°C
Color White-Yellow
Boiling Point 136°C
Quantity 5 g
Formula Weight 234.04
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodobenzyl Alcohol
RTECSNumber DO8318020
TSCA No