accessibility menu, dialog, popup

UserName

HOI

CAS RN 18282-51-4

IUPAC Name: (4-iodophenyl)methanol
Synonyms: benzenemethanol, 4-iodo-
Molecular Weight (g/mol): 234.04
Molecular Formula: C7H7IO
InChi Key: CNQRHSZYVFYOIE-UHFFFAOYSA-N
SMILES: OCC1=CC=C(I)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)