missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzyl alcohol, 97%

CAS: 18282-51-4 | C7H7IO | 234.036 g/mol

Supplier:  Thermo Scientific Chemicals L1927806

 View more versions of this product

Catalog No. AAL1927806


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Iodobenzyl alcohol
72°C to 74°C
MFCD01732720
1931621
4-iodobenzyl alcohol, 4-iodophenyl methanol, p-iodobenzyl alcohol, benzenemethanol, 4-iodo, benzyl alcohol, p-iodo, 4-iodobenzylalcohol, p-iodobenzylalcohol, 4-iodobenzyl-alcohol, 4-iodobenzyl zlcohol, p-lodo-benzyl alcohol
C1=CC(=CC=C1CO)I
234.036
234.04
18282-51-4
C7H7IO
5 g
Light sensitive
CNQRHSZYVFYOIE-UHFFFAOYSA-N
(4-iodophenyl)methanol
29012
97%

Chemical Identifiers

18282-51-4
234.036
CNQRHSZYVFYOIE-UHFFFAOYSA-N
29012
C1=CC(=CC=C1CO)I
C7H7IO
MFCD01732720
4-iodobenzyl alcohol, 4-iodophenyl methanol, p-iodobenzyl alcohol, benzenemethanol, 4-iodo, benzyl alcohol, p-iodo, 4-iodobenzylalcohol, p-iodobenzylalcohol, 4-iodobenzyl-alcohol, 4-iodobenzyl zlcohol, p-lodo-benzyl alcohol
(4-iodophenyl)methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only