missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzyl alcohol, 97%

Catalog No. AAL1927803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1927803 1 g
AAL1927806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1927803 Supplier Thermo Scientific Chemicals Supplier No. L1927803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18282-51-4 | C7H7IO | 234.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18282-51-4
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD01732720
InChI Key CNQRHSZYVFYOIE-UHFFFAOYSA-N
Synonym 4-iodobenzyl alcohol, 4-iodophenyl methanol, p-iodobenzyl alcohol, benzenemethanol, 4-iodo, benzyl alcohol, p-iodo, 4-iodobenzylalcohol, p-iodobenzylalcohol, 4-iodobenzyl-alcohol, 4-iodobenzyl zlcohol, p-lodo-benzyl alcohol
PubChem CID 29012
IUPAC Name (4-iodophenyl)methanol
SMILES C1=CC(=CC=C1CO)I

Specifications

Melting Point 72°C to 74°C
Quantity 1 g
Beilstein 1931621
Sensitivity Light sensitive
Formula Weight 234.04
Percent Purity 97%
Chemical Name or Material 4-Iodobenzyl alcohol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only