missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodobenzotrifluoride (stabilized with Copper chip) 97.0+%, TCI America™
$113.71 - $335.52
Chemical Identifiers
| CAS | 455-13-0 |
|---|---|
| Molecular Formula | C7H4F3I |
| Molecular Weight (g/mol) | 272.009 |
| MDL Number | MFCD00039398 |
| InChI Key | SKGRFPGOGCHDPC-UHFFFAOYSA-N |
| Synonym | 4-iodobenzotrifluoride, 1-iodo-4-trifluoromethyl benzene, p-iodobenzotrifluoride, benzene, 1-iodo-4-trifluoromethyl, 4-trifluoromethyl iodobenzene, 4-iodo-alpha,alpha,alpha-trifluorotoluene, alpha,alpha,alpha-trifluoro-4-iodotoluene, 4-iodobenztrifluoride, 4-iodobenzo trifluoride, 4-iodo-1-trifluoromethylbenzene |
| PubChem CID | 67993 |
| IUPAC Name | 1-iodo-4-(trifluoromethyl)benzene |
| SMILES | C1=CC(=CC=C1C(F)(F)F)I |
Chemical Identifiers
| 455-13-0 | |
| 272.009 | |
| SKGRFPGOGCHDPC-UHFFFAOYSA-N | |
| 67993 | |
| C1=CC(=CC=C1C(F)(F)F)I |
| C7H4F3I | |
| MFCD00039398 | |
| 4-iodobenzotrifluoride, 1-iodo-4-trifluoromethyl benzene, p-iodobenzotrifluoride, benzene, 1-iodo-4-trifluoromethyl, 4-trifluoromethyl iodobenzene, 4-iodo-alpha,alpha,alpha-trifluorotoluene, alpha,alpha,alpha-trifluoro-4-iodotoluene, 4-iodobenztrifluoride, 4-iodobenzo trifluoride, 4-iodo-1-trifluoromethylbenzene | |
| 1-iodo-4-(trifluoromethyl)benzene |
Specifications
| 455-13-0 | |
| 185°C | |
| MFCD00039398 | |
| 4-iodobenzotrifluoride, 1-iodo-4-trifluoromethyl benzene, p-iodobenzotrifluoride, benzene, 1-iodo-4-trifluoromethyl, 4-trifluoromethyl iodobenzene, 4-iodo-alpha,alpha,alpha-trifluorotoluene, alpha,alpha,alpha-trifluoro-4-iodotoluene, 4-iodobenztrifluoride, 4-iodobenzo trifluoride, 4-iodo-1-trifluoromethylbenzene | |
| C1=CC(=CC=C1C(F)(F)F)I | |
| 272.009 | |
| 272.01 | |
| Liquid |
| Yellow | |
| C7H4F3I | |
| 5 g | |
| SKGRFPGOGCHDPC-UHFFFAOYSA-N | |
| 1-iodo-4-(trifluoromethyl)benzene | |
| 67993 | |
| ≥97.0% (GC) | |
| 4-Iodobenzotrifluoride (stabilized with Copper chip) |
Safety and Handling
TSCA : Yes