missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzotrifluoride (stabilized with Copper chip) 97.0+%, TCI America™

Catalog No. i05505g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
I05505G 5 g
I055025G 25 g
2 options

Catalog No. I05505G

Supplier: TCI America I05505G

Only null left

Chemical Identifiers

CAS 455-13-0
Molecular Formula C7H4F3I
Molecular Weight (g/mol) 272.009
MDL Number MFCD00039398
InChI Key SKGRFPGOGCHDPC-UHFFFAOYSA-N
Synonym 4-iodobenzotrifluoride, 1-iodo-4-trifluoromethyl benzene, p-iodobenzotrifluoride, benzene, 1-iodo-4-trifluoromethyl, 4-trifluoromethyl iodobenzene, 4-iodo-alpha,alpha,alpha-trifluorotoluene, alpha,alpha,alpha-trifluoro-4-iodotoluene, 4-iodobenztrifluoride, 4-iodobenzo trifluoride, 4-iodo-1-trifluoromethylbenzene
PubChem CID 67993
IUPAC Name 1-iodo-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)I

Specifications

Color Yellow
Boiling Point 185°C
Quantity 5 g
Formula Weight 272.01
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Iodobenzotrifluoride (stabilized with Copper chip)