missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodobenzonitrile 98.0+%, TCI America™
Supplier: TCI America I06615G
Specifications
| 4-Iodobenzonitrile | |
| 127°C | |
| C7H4IN | |
| 5 g | |
| p-iodobenzonitrile, benzonitrile, 4-iodo, p-cyanoiodobenzene, benzonitrile, p-iodo, 1-cyano-4-iodobenzene, 4-cyanoiodobenzene, 4-iodo-benzonitrile, 4-iodobenzenecarbonitrile, 4-iodbenzonitril, 4-iodo benzonitrile | |
| IC1=CC=C(C=C1)C#N | |
| 229.02 | |
| 229.02 | |
| Crystalline Powder |
| 3058-39-7 | |
| White-Yellow | |
| MFCD00051310 | |
| 3439 | |
| XOKDXPVXJWTSRM-UHFFFAOYSA-N | |
| 4-iodobenzonitrile | |
| 76467 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3058-39-7 | |
| 229.02 | |
| XOKDXPVXJWTSRM-UHFFFAOYSA-N | |
| 76467 | |
| IC1=CC=C(C=C1)C#N |
| C7H4IN | |
| MFCD00051310 | |
| p-iodobenzonitrile, benzonitrile, 4-iodo, p-cyanoiodobenzene, benzonitrile, p-iodo, 1-cyano-4-iodobenzene, 4-cyanoiodobenzene, 4-iodo-benzonitrile, 4-iodobenzenecarbonitrile, 4-iodbenzonitril, 4-iodo benzonitrile | |
| 4-iodobenzonitrile |
Safety and Handling
TSCA : Yes