missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzonitrile 98.0+%, TCI America™

Catalog No. p-7131528
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
I06615G 5 g
I066125G 25 g
2 options

Catalog No. I06615G

Supplier: TCI America I06615G

Only null left

Chemical Identifiers

CAS 3058-39-7
Molecular Formula C7H4IN
Molecular Weight (g/mol) 229.02
MDL Number MFCD00051310
InChI Key XOKDXPVXJWTSRM-UHFFFAOYSA-N
Synonym p-iodobenzonitrile, benzonitrile, 4-iodo, p-cyanoiodobenzene, benzonitrile, p-iodo, 1-cyano-4-iodobenzene, 4-cyanoiodobenzene, 4-iodo-benzonitrile, 4-iodobenzenecarbonitrile, 4-iodbenzonitril, 4-iodo benzonitrile
PubChem CID 76467
IUPAC Name 4-iodobenzonitrile
SMILES IC1=CC=C(C=C1)C#N

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 229.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodobenzonitrile