Learn More
4-Iodoanisole, 98+%
CAS: 696-62-8 | C7H7IO | 234.036 g/mol
Supplier: Thermo Scientific Chemicals A1214422
Description
4-Iodoanisole is used in wide range of medicals industrial applications as well as in human and animal nutrition products, pharmaceutical intermediates, polarizing films for Liquid Crystal Display (LCD) chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Iodoanisole is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents.
Solubility
Soluble in ethanol, ether, chloroform.
Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.
Specifications
| 4-Iodoanisole | |
| 48°C to 52°C | |
| >110°C (230°F) | |
| MFCD00001056 | |
| 1906692 | |
| 4-iodoanisole, p-iodoanisole, benzene, 1-iodo-4-methoxy, isoform, 4-methoxyiodobenzene, p-methoxyiodobenzene, anisole, p-iodo, 4-iodomethoxybenzene, p-iodophenyl methyl ether, 1-iodo-4-methoxy-benzene | |
| SYSZENVIJHPFNL-UHFFFAOYSA-N | |
| 1-iodo-4-methoxybenzene | |
| 69676 | |
| ≥98% |
| 696-62-8 | |
| 240°C | |
| C7H7IO | |
| 100 g | |
| Light sensitive | |
| Soluble in ethanol,ether,chloroform. | |
| COC1=CC=C(C=C1)I | |
| 234.036 | |
| 234.04 |
Chemical Identifiers
| 696-62-8 | |
| 234.036 | |
| SYSZENVIJHPFNL-UHFFFAOYSA-N | |
| 69676 | |
| COC1=CC=C(C=C1)I |
| C7H7IO | |
| MFCD00001056 | |
| 4-iodoanisole, p-iodoanisole, benzene, 1-iodo-4-methoxy, isoform, 4-methoxyiodobenzene, p-methoxyiodobenzene, anisole, p-iodo, 4-iodomethoxybenzene, p-iodophenyl methyl ether, 1-iodo-4-methoxy-benzene | |
| 1-iodo-4-methoxybenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 211-798-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only