missing translation for 'onlineSavingsMsg'
Learn More

4-Iodoanisole, 98+%

Catalog No. p-7081947
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1214414 25 g
AAA1214422 100 g
AAA1214436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1214414 Supplier Thermo Scientific Chemicals Supplier No. A1214414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 696-62-8 | C7H7IO | 234.036 g/mol

4-Iodoanisole is used in wide range of medicals industrial applications as well as in human and animal nutrition products, pharmaceutical intermediates, polarizing films for Liquid Crystal Display (LCD) chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodoanisole is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents.

Solubility
Soluble in ethanol, ether, chloroform.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.

Chemical Identifiers

CAS 696-62-8
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD00001056
InChI Key SYSZENVIJHPFNL-UHFFFAOYSA-N
Synonym 4-iodoanisole, p-iodoanisole, benzene, 1-iodo-4-methoxy, isoform, 4-methoxyiodobenzene, p-methoxyiodobenzene, anisole, p-iodo, 4-iodomethoxybenzene, p-iodophenyl methyl ether, 1-iodo-4-methoxy-benzene
PubChem CID 69676
IUPAC Name 1-iodo-4-methoxybenzene
SMILES COC1=CC=C(C=C1)I

Specifications

Melting Point 48°C to 52°C
Boiling Point 240°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1906692
Sensitivity Light sensitive
Solubility Information Soluble in ethanol,ether,chloroform.
Formula Weight 234.04
Percent Purity ≥98%
Chemical Name or Material 4-Iodoanisole
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 211-798-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only