missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodo-m-xylene 98.0+%, TCI America™
Supplier: TCI America I056125G
Specifications
| 4-Iodo-m-xylene | |
| Yellow | |
| 25 g | |
| C8H9I | |
| 4-iodo-m-xylene, 2,4-dimethyliodobenzene, 1,3-dimethyl-4-iodobenzene, m-xylene, 4-iodo, benzene, 1-iodo-2,4-dimethyl, 2,4-dimethyl-1-iodobenzene, 2-iodo-1,5-dimethylbenzene, 4-jod-m-xylol, pubchem3869, acmc-209jnr | |
| CC1=CC=C(I)C(C)=C1 | |
| 232.06 | |
| 232.06 |
| 4214-28-2 | |
| 233°C | |
| Clear Liquid at 20°C | |
| MFCD00013706 | |
| BUNKQJAMHYKQIM-UHFFFAOYSA-N | |
| 1-iodo-2,4-dimethylbenzene | |
| 77885 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 4214-28-2 | |
| 232.06 | |
| BUNKQJAMHYKQIM-UHFFFAOYSA-N | |
| 77885 | |
| CC1=CC=C(I)C(C)=C1 |
| C8H9I | |
| MFCD00013706 | |
| 4-iodo-m-xylene, 2,4-dimethyliodobenzene, 1,3-dimethyl-4-iodobenzene, m-xylene, 4-iodo, benzene, 1-iodo-2,4-dimethyl, 2,4-dimethyl-1-iodobenzene, 2-iodo-1,5-dimethylbenzene, 4-jod-m-xylol, pubchem3869, acmc-209jnr | |
| 1-iodo-2,4-dimethylbenzene |
Safety and Handling
TSCA : Yes