missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-m-xylene 98.0+%, TCI America™

Catalog No. p-7131484
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05615G 5 g
I056125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05615G Supplier TCI America Supplier No. I05615G
Only null left

Chemical Identifiers

CAS 4214-28-2
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.06
MDL Number MFCD00013706
InChI Key BUNKQJAMHYKQIM-UHFFFAOYSA-N
Synonym 4-iodo-m-xylene, 2,4-dimethyliodobenzene, 1,3-dimethyl-4-iodobenzene, m-xylene, 4-iodo, benzene, 1-iodo-2,4-dimethyl, 2,4-dimethyl-1-iodobenzene, 2-iodo-1,5-dimethylbenzene, 4-jod-m-xylol, pubchem3869, acmc-209jnr
PubChem CID 77885
IUPAC Name 1-iodo-2,4-dimethylbenzene
SMILES CC1=CC=C(I)C(C)=C1

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 233°C
Formula Weight 232.06
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Iodo-m-xylene