missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-4'-nitrobiphenyl 98.0+%, TCI America™

Catalog No. I05371G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05371G 1 g
I05375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05371G Supplier TCI America Supplier No. I05371G

Chemical Identifiers

CAS 29170-08-9
Molecular Formula C12H8INO2
Molecular Weight (g/mol) 325.11
MDL Number MFCD00024698
InChI Key YEDVRRNLTGKJSW-UHFFFAOYSA-N
PubChem CID 120142
IUPAC Name 4-iodo-4'-nitro-1,1'-biphenyl
SMILES [O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(I)C=C1

Specifications

Melting Point 216°C
Color Yellow
Quantity 1 g
Formula Weight 325.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-4′-nitrobiphenyl