accessibility menu, dialog, popup

UserName

CAS RN 29170-08-9

ONOI

CAS RN 29170-08-9

IUPAC Name: 4-iodo-4'-nitro-1,1'-biphenyl
Synonyms: 4-iodo-4'-nitro-1,1'-biphenyl
Molecular Weight (g/mol): 325.11
Molecular Formula: C12H8INO2
InChi Key: YEDVRRNLTGKJSW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(I)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results