missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2-(trifluoromethyl)benzonitrile 98.0+%, TCI America™

Catalog No. p-7131718
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10181G 1 g
I10185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10181G Supplier TCI America Supplier No. I10181G
Only null left

Chemical Identifiers

CAS 101066-87-9
Molecular Formula C8H3F3IN
Molecular Weight (g/mol) 297.019
MDL Number MFCD08458091
InChI Key RBRTXKYFMXVIER-UHFFFAOYSA-N
Synonym 4-iodo-2-trifluoromethyl benzonitrile, 2-cyano-5-iodobenzotrifluoride, benzonitrile, 4-iodo-2-trifluoromethyl, 2-trifluoromethyl-4-iodobenzonitrile, 4-cyano-3-trifluoromethyl-1-iodobenzene, 4-cyano-3-trifluoromethyl iodobenzene, 4-iodo-2-trifluoromethyl benzenecarbonitrile, pubchem4810, acmc-1bqw7, ksc494m4b
PubChem CID 16218337
IUPAC Name 4-iodo-2-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1I)C(F)(F)F)C#N

Specifications

Melting Point 54°C
Quantity 1 g
UN Number 3439
Formula Weight 297.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-2-(trifluoromethyl)benzonitrile