accessibility menu, dialog, popup

UserName

CAS RN 101066-87-9

IUPAC Name: 4-iodo-2-(trifluoromethyl)benzonitrile
Synonyms: 4-iodo-2-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 297.02
Molecular Formula: C8H3F3IN
InChi Key: RBRTXKYFMXVIER-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(C=CC(I)=C1)C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)