missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2-nitrotoluene, 97%

Catalog No. AAA1860706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1860706 5 g
AAA1860714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1860706 Supplier Thermo Scientific Chemicals Supplier No. A1860706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 41252-97-5 | C7H6INO2 | 263.034 g/mol

4-Iodo-2-nitrotoluene is used to produce 3-(4-methyl-3-nitro-phenyl)-1H-indole at the temperature of 40°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-2-nitrotoluene is used to produce 3-(4-methyl-3-nitro-phenyl)-1H-indole at the temperature of 40°C.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.

Chemical Identifiers

CAS 41252-97-5
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.034
MDL Number MFCD00051090
InChI Key QLMRDNPXYNJQMQ-UHFFFAOYSA-N
Synonym 4-iodo-2-nitrotoluene, benzene, 4-iodo-1-methyl-2-nitro, pubchem15938, 2-nitro-4-iodotoluene, acmc-1apfk, maybridge1_008109, ksc493s9d, 4-iodo-1-methyl-2-nitro-benzene
PubChem CID 170481
IUPAC Name 4-iodo-1-methyl-2-nitrobenzene
SMILES CC1=C(C=C(C=C1)I)[N+](=O)[O-]

Specifications

Melting Point 58°C to 62°C
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 263.03
Percent Purity 97%
Chemical Name or Material 4-Iodo-2-nitrotoluene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 255-279-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only