accessibility menu, dialog, popup

UserName

CAS RN 41252-97-5

IUPAC Name: 4-iodo-1-methyl-2-nitrobenzene
Synonyms: 4-iodo-1-methyl-2-nitrobenzene
Molecular Weight (g/mol): 263.03
Molecular Formula: C7H6INO2
InChi Key: QLMRDNPXYNJQMQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(I)C=C1[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)