missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylacetone 97.0+%, TCI America™

Catalog No. p-7130937
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12495G 5 g
H124925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12495G Supplier TCI America Supplier No. H12495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 770-39-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00210439
InChI Key VWMVAQHMFFZQGD-UHFFFAOYSA-N
Synonym 4-hydroxyphenylacetone, 1-4-hydroxyphenyl propan-2-one, 1-4-hydroxyphenyl acetone, p-hydroxyphenylacetone, 1-4-hydroxyphenyl-2-propanone, 2-propanone, 1-4-hydroxyphenyl, 4-hydroxyphenyl acetone, 4-hydroxyphenylpropanone, acmc-209p7o, 4-hydroxyphenylpropan-2-one
PubChem CID 7019274
IUPAC Name 1-(4-hydroxyphenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(O)C=C1

Specifications

Melting Point 40°C
Color White-Yellow
Boiling Point 177°C
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Hydroxyphenylacetone