accessibility menu, dialog, popup

UserName

CAS RN 770-39-8

IUPAC Name: 1-(4-hydroxyphenyl)propan-2-one
Synonyms: p-hydroxyphenyl-1-propanone
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: VWMVAQHMFFZQGD-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results