missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylacetic Acid 99.0+%, TCI America™

Catalog No. H029025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H029025G 25 g
H0290100G 100 g
H0290500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H029025G Supplier TCI America Supplier No. H029025G

Chemical Identifiers

CAS 156-38-7
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00004347
InChI Key XQXPVVBIMDBYFF-UHFFFAOYSA-N
Synonym 4-hydroxyphenylacetic acid, 2-4-hydroxyphenyl acetic acid, p-hydroxyphenylacetic acid, 4-hydroxyphenyl acetic acid, 4-hydroxyphenylacetate, 4-hydroxybenzeneacetic acid, benzeneacetic acid, 4-hydroxy, p-hydroxyphenyl acetic acid, parahydroxy phenylacetic acid, 4-carboxymethylphenol
PubChem CID 127
ChEBI CHEBI:18101
IUPAC Name 2-(4-hydroxyphenyl)acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)O

Specifications

Melting Point 150°C
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxyphenylacetic Acid
EINECSNumber (9)-1022
RTECSNumber AI2680000
TSCA Yes