missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenoxyacetic acid, 98+%

Catalog No. AAA1294006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1294006 5 g
AAA1294014 25 g
AAA1294022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1294006 Supplier Thermo Scientific Chemicals Supplier No. A1294006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1878-84-8 | C8H8O4 | 168.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1878-84-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00014362
InChI Key PKGWLCZTTHWKIZ-UHFFFAOYSA-N
Synonym 4-hydroxyphenoxyacetic acid, 4-hydroxyphenoxy acetic acid, 2-4-hydroxyphenoxy acetic acid, 4-hydroxypheoxyacetate, acetic acid, 4-hydroxyphenoxy, 4-hydroxy-phenoxy-acetic acid, p-hydroxyphenoxyacetic acid, acetic acid, 2-4-hydroxyphenoxy, acetic acid, p-hydroxyphenoxy, 4-carboxymethoxy phenol
PubChem CID 15881
ChEBI CHEBI:1881
IUPAC Name 2-(4-hydroxyphenoxy)acetic acid
SMILES C1=CC(=CC=C1O)OCC(=O)O

Specifications

Melting Point 153°C to 158°C
Quantity 5 g
Beilstein 1952779
Formula Weight 168.15
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxyphenoxyacetic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 217-525-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only