missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Hydroxymethyl)phenylacetic Acid 97.0+%, TCI America™
$106.25 - $311.30
Chemical Identifiers
| CAS | 73401-74-8 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.176 |
| MDL Number | MFCD00065692 |
| InChI Key | FWZBPBKAANKOJQ-UHFFFAOYSA-N |
| PubChem CID | 3283498 |
| IUPAC Name | 2-[4-(hydroxymethyl)phenyl]acetic acid |
| SMILES | C1=CC(=CC=C1CC(=O)O)CO |
Chemical Identifiers
| 73401-74-8 | |
| 166.176 | |
| FWZBPBKAANKOJQ-UHFFFAOYSA-N | |
| 2-[4-(hydroxymethyl)phenyl]acetic acid |
| C9H10O3 | |
| MFCD00065692 | |
| 3283498 | |
| C1=CC(=CC=C1CC(=O)O)CO |
Specifications
| 73401-74-8 | |
| White | |
| MFCD00065692 | |
| 4-hydroxymethyl phenylacetic acid, 2-4-hydroxymethyl phenyl acetic acid, 4-hydroxymethyl phenyl acetic acid, 4-hydroxymethylphenylacetic acid, benzeneacetic acid,4-hydroxymethyl, benzeneacetic acid, 4-hydroxymethyl, ambotzrl-1037, pubchem19630, acmc-1bl5i | |
| C1=CC(=CC=C1CC(=O)O)CO | |
| 166.176 | |
| 166.18 | |
| Crystalline Powder |
| 130°C | |
| C9H10O3 | |
| 1 g | |
| FWZBPBKAANKOJQ-UHFFFAOYSA-N | |
| 2-[4-(hydroxymethyl)phenyl]acetic acid | |
| 3283498 | |
| ≥97.0% (T) | |
| 4-(Hydroxymethyl)phenylacetic Acid |
Safety and Handling
TSCA : No