accessibility menu, dialog, popup

UserName

OOHOH

CAS RN 73401-74-8

IUPAC Name: 2-[4-(hydroxymethyl)phenyl]acetic acid
Synonyms: [4-(hydroxymethyl)phenyl]acetic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: FWZBPBKAANKOJQ-UHFFFAOYSA-N
SMILES: OCC1=CC=C(CC(O)=O)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)