missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)phenylacetic Acid 97.0+%, TCI America™

Catalog No. H14731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14731G 1 g
H14735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14731G Supplier TCI America Supplier No. H14731G

Chemical Identifiers

CAS 73401-74-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00065692
InChI Key FWZBPBKAANKOJQ-UHFFFAOYSA-N
PubChem CID 3283498
IUPAC Name 2-[4-(hydroxymethyl)phenyl]acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)CO

Specifications

Melting Point 130°C
Color White
Quantity 1 g
Synonym 4-hydroxymethyl phenylacetic acid, 2-4-hydroxymethyl phenyl acetic acid, 4-hydroxymethyl phenyl acetic acid, 4-hydroxymethylphenylacetic acid, benzeneacetic acid,4-hydroxymethyl, benzeneacetic acid, 4-hydroxymethyl, ambotzrl-1037, pubchem19630, acmc-1bl5i
Formula Weight 166.18
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Hydroxymethyl)phenylacetic Acid